Xylazine Test Strip with Micro Scoop, WiseBatch
Supplier: WISEBATCH LLC MS
XYL-WB2
MSPP-XYL-WB2PK 130 USD
MSPP-XYL-WB2
Xylazine Test Strip with Micro Scoop, WiseBatch
Drug Tests
The WiseBatch Xylazine Test Strip has the ability to test for trace amounts of Xylazine in drug samples.
  • Testing sensitivity to a concentration as low as 500 ng/ml
  • Results are displayed within 1 to 2 minutes
  • 99% + accuracy rating
  • 18 to 24 month shelf life
The test strip includes a micro scoop, simple instructions for use, as well as a scannable QR code that leads to the resources section on the WiseBatch website where users can find a broad spectrum of testing education.
This product must be stored at 2 to 30° C (35.6 to 86° F).
Ordering information: Includes micro scoop.
Packaging: Includes 1 pack of 100 individually wrapped xylazine test strips.