96-Well Polypropylene Storage System, Costar, Cole-Parmer
Supplier: Cole Parmer
01728-80
MSPP-VV0172880CS 503.2 USD
MSPP-VV0172880
96-Well Polypropylene Storage System, Costar, Cole-Parmer
Microplates Deep Well Microplates
Standard microplates are built to industry standard footprint, compatible with automated systems.
  • Certified RNase- and Dnase-free
Constructed of clarified polypropylene for optimal clarity and solvent resistance which is ideal for compound library and combinatorial chemistry applications.