Cooling/heating Recirculating Fluid Block, 120 V
Supplier: V & P SCIENTIFIC INC MS
VP744A-1
MSPP-VP744A-1EA 3031.38 USD
MSPP-VP744A-1
Cooling/heating Recirculating Fluid Block, 120 V
Block Heaters Block Heater/Coolers
Heater/chiller, recirculating fluid block, SLAS microplate pocket on top, anodized aluminum, SLAS locating feature on the bottom, inlet/outlet ³/₁₆ inch tubing fittings on short side.
  • V&P Scientific builds robust, low-profile, ambient to 200 °C heat blocks that enable heating and stirring in standard microplates, deep well microplates, or reaction blocks of glass tubes and sealed vials
  • constructed out of a specially designed Eddy Current defeating aluminum designed to be used with our strong NdFeB tumble stirrers
Each heat block comes with one orMore Product Information