Heating Block, Deep Chamber, 120 V
Supplier: V & P SCIENTIFIC INC MS
VP741D
MSPP-VP741DEA 8946 USD
MSPP-VP741D
Heating Block, Deep Chamber, 120 V
Block Heaters Block Heater/Coolers
HEAT BLOCK, deep chamber for microplates or racks of tubes/vials, designed to heat and stir, anodized aluminum, PID controller, 12.5 mm Mica SLAS adaptor on bottom.