Vertical Tumble Stirrer, Double Stack Servo Motor, 5 SLAS Positions
Supplier: V & P SCIENTIFIC INC MS
VP710E5X
MSPP-VP710E5XEA 20187.62 USD
MSPP-VP710E5X
Vertical Tumble Stirrer, Double Stack Servo Motor, 5 SLAS Positions
Stirrers Magnetic Stirrers
The Alligator Magnetic Vertical Tumble Stirrer uses patented stir-cylinder technology to mix large numbers of samples in tubes, vials, bottles, and microplates. Instead of spinning a magnetic stir bar about the horizontal plane like the standard magnetic flask stirrer, the Alligator Vertical Tumble Stirrer causes stir elements of all sizes and shapes to tumble vertically end-over-end inside each well or vessel. The Alligator Vertical Tumble Stirrer will stir all types of microplates of any volume or format. It will also stir V and U bottom microplates, PCR plates, micro-centrifuge tubes, test tubes, syringes, vials and bottles, and specially designed troughs and reservoirs.
  • Can stir in any vessel regardless of the size or shape
  • No cross contamination – wells do not have to be sealed
  • Simple to operate, full-speed control, computer control available
  • Can set exact rpm for reproducible results day to day
  • Designed to be compatible with most robotic systems
  • Flexibility of stir deck alignment, durable – can stir continuously for years without maintenance
High–throughput stirring in stacks of microplates can increase the aeration of microbial cultures and increase yield and run multiple experiments in parallel, complete stirring of large numbers of samples. it can stir chemical reactions to speed completion and stir and heat to 200 °C at the same time. Get more reactions per gram of starting material.
Using the most powerful permanent magnets manufactured, we make many different models of Vertical Tumble Stirrers that fit the many varied applications and requirements of our customers. Requirements for heating and continuous stirring for a long period of time demand special Vertical Tumble Stirrers that are resistant to heat and with motors capable of working under high loads for extended periods. We make our Vertical Tumble Stirrers heat resistant using Mica. It will stir even 100000 centistoke viscous solutions (6.6 times more viscous than honey), even thick slurries and emulsions.