VacciGrade™ Pam3CSK4, Triacylated Lipopeptide
Supplier: Invivogen
VAC-PMS
MSPP-VACPMSEA 550.77 USD
MSPP-VACPMS
VacciGrade™ Pam3CSK4, Triacylated Lipopeptide
Proteins and Peptides
This is a synthetic lipopeptide and a TLR2/1 ligand.
  • As an adjuvant, it has improved the efficacy of a DNA-based vaccine against Leishmania
It has been reported to efficiently increase antibody responses to influenza antigens and to exert a strong local response. It is prepared under strict aseptic conditions, is guaranteed sterile and is thoroughly tested to confirm the absence of endotoxins.
Caution: For research use only. Not intended for use in humans.