Aluminium hydroxide, Alhydrogel® adjuvant 2% wet gel suspension

Supplier: Invivogen

VAC-ALU-250
MSPP-VACALU250EA 595.38 USD
MSPP-VACALU250
Aluminium hydroxide, Alhydrogel® adjuvant 2% wet gel suspension
Aluminium hydroxide

Alhydrogel® adjuvant is an aluminium hydroxide (referred to as alum) wet gel suspension.


Alhydrogel® adjuvant 2% is provided as a ready-to-use, aluminium hydroxide wet gel (colloidal) suspension. Alhydrogel® adjuvant is sterilized by autoclavation and aseptically filled.

Formula: Al(OH)₃
MDL Number: MFCD00003420
CAS Number: 21645-51-2

Order Now

Specification Test Results

Appearance White Gelatinous Precipitate
Aluminium Content 9.0 - 11.0 mg/ml
pH ∼6.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR