Nalgene® Carboys, LDPE, with Handles and Spigot, Thermo Scientific
Supplier: Cole Parmer
06065-32
MSPP-UX0606532EA 395.2 USD
MSPP-UX0606532
Nalgene® Carboys, LDPE, with Handles and Spigot, Thermo Scientific
Bottles Carboys
Round carboys made of LDPE with handles and spigot.
  • Polypropylene (PP) cap ensures leakproof seal
  • Spigots accept ⁵/₈" Int.Ø tubing
Delivery information: Supplied with adapters for ¼ and ½" Int.Ø tubing.