Gaymar Heating/Cooling System T/Pump, Braintree Scientific
Supplier: BRAINTREE SCIENTIFIC INC MS
TP-700
MSPP-TP700EA 1092.31 USD
MSPP-TP700
Gaymar Heating/Cooling System T/Pump, Braintree Scientific
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
The T/Pump® system provides safe and effective localized warming and cooling therapy with precise temperature control.
  • Convenient operation with tap water
  • Ergonomic design
  • Audible and visible alarms alert
  • Large transluscent reservoir
  • Three-layer safety system
  • Multiple temperature settings and treatment cycles
  • Large buttons and LED indicators
Simple and versatile, T/Pump is effective for chronic pain relief and patient comfort in orthopedic conditions, skin trauma and a number of other applications to meet the needs of an ever-changing patient population. The large translucent reservoir and large buttons make operation simple.