Jess or Wes Separation Module, Protein Simple
Supplier: 0000014126
SM-W002
MSPP-SMW002EA 1395.38 USD
MSPP-SMW002
Jess or Wes Separation Module, Protein Simple
Assays Protein Assays
Reagents for 104 data points to use with a detection module.
It includes two parts: Separation box with 8 capillary Cartridges (13 capillaries per cartridge), 8 Jess/Wes 12 to 230 kDa pre-filled Plates (pre-filled with separation matrix 2, stacking matrix 2, split running buffer 2 and matrix removal buffer), sample buffer, wash buffer, plus standard pack 1 clamshell with 8 ea. DTT, FL standard master mix with 29 kDa system control protein.