Human FcgBP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEP389HU
MSPP-SEP389HUEA 770 USD
MSPP-SEP389HU
Human FcgBP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human FcgBP (Fc Fragment Of IgG Binding Protein). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.117 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Cloning a cDNA for human IgGFc binding protein (FcgammaBP) from human colonic epithelial cells reveals an mRNA and coding region of 17 and 16.2 kilobases, respectively. The predicted amino acid sequence contains 12 occurrences of a 400-amino acid cysteine-rich unit resembling that found in mucin. A motif (CGLCGN) in FcgammaBP is conserved in MUC2 and prepro-von Willebrand factor. The N-terminal 450-amino acid sequences are necessary and sufficient to confer IgG Fc binding activity. FcgammaBP mRNA is expressed only in placenta and colonic epithelial cells. These results suggest that FcgammaBP may play an important role in immune protection and inflammation in the intestines of primates.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.