Rat Rac1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEM427RA
MSPP-SEM427RAEA 836 USD
MSPP-SEM427RA
Rat Rac1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat Rac1 (Ras Related C3 Botulinum Toxin Substrate 1). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 32 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Rac1 is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene. This mutant did bind to POR1 and it induced membrane ruffling and transformation. A second RAC mutant, which bound PAK but not POR1, induced JNK activation but was defective in inducing membrane ruffling and transformation. The authors concluded that the effects of RAC on the JNK cascade and on actin polymerization and cell proliferation are mediated by distinct effector functions that diverge at the level of RAC itself.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.