Rat NFE2L2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEL947RA
MSPP-SEL947RAEA 836 USD
MSPP-SEL947RA
Rat NFE2L2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat NFE2L2 (Nuclear Factor Erythroid Derived 2 Like Protein 2). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.057 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Nrf2 is a master regulator of the antioxidant response. The antioxidant response is important for the amelioration of oxidative stress. Oxidative stress can result in cancer, cardiovascular diseases, inflammation, neurological diseases, and renal disease. Because Nrf2 is able to induce genes important in combating oxidative stress, thereby activating the body’s own protective response, it is able to protect from a variety of [oxidative stress]-related complications, even in situations where the administration of exogenous antioxidants have failed. NFE2, NFE2L1, and NFE2L2 comprise a family of human genes encoding basic leucine zipper (bZIP) transcription factors. They share highly conserved regions that are distinct from other bZIP families, such as JUN and FOS, although remaining regions have diverged considerably from each other.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.