Human SEMA3C ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEL919HU
MSPP-SEL919HUEA 770 USD
MSPP-SEL919HU
Human SEMA3C ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human SEMA3C (Semaphorin 3C). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 27 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
The 616-amino acid human semaphorin E contains an N-terminal signal sequence of 15 amino acids that is followed by a semaphorin domain. in vitro translation of the semaphorin E cDNA yielded a 70 kDa protein. The authors showed that semaphorin E is secreted by mammalian cells. By Northern blot analysis, semaphorin E was expressed as a 5.2-kb transcript in all human tissues tested. CDDP-sensitive cells transfected with semaphorin E cDNA acquired CDDP-resistance. Semaphorin E was overexpressed in CDDP-resistant cell lines and was induced by diverse chemotherapeutic drugs and by x-ray and UV irradiation. Aberrant semaphorin E protein expression was detected immunohistochemically
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.