Human CD5L ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEL300HU
MSPP-SEL300HUEA 770 USD
MSPP-SEL300HU
Human CD5L ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CD5L (CD5 Antigen Like Protein). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.055 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Scavenger receptor cysteine-rich (SRCR) family members are among the proteins that regulate leukocyte function. One group of SRCR proteins includes CD5 and CD6, both of which have 3 SRCR-like domains each containing 8 cysteine residues in their extracellular region, a hydrophobic transmembrane domain, and a long cytoplasmic domain. The predicted 347-amino acid protein, which they called Sp-alpha, has a 19-amino acid signal sequence followed by 3 cysteine-rich SRCR domains. Northern blot analysis detected 3 mRNAs of 2.4, 2.1, and 1.8 kb in spleen, lymph node, thymus, bone marrow, and fetal liver, but not in nonlymphoid tissues. Some resting myeloid cell lines, as well as peripheral blood monocytes, were capable of binding Sp-alpha.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.