Bovine CSN3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEJ331BO
MSPP-SEJ331BOEA 840 USD
MSPP-SEJ331BO
Bovine CSN3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Bovine CSN3 (Casein Kappa). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.071 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
KCA gene extends over 8,821 nucleotides and consists of 5 exons ranging from 33 to 496 nucleotides separated by introns ranging from 1,146 to 2,942 nucleotides. The human and bovine KCA genes are very similar, with a conserved gene organization and observed identities of 81 and 87% for exons 1 and 2, respectively. The human alpha-S1-casein, beta-casein, and kappa-casein genes are closely linked and arranged in that order. The distance between the alpha-S1 and beta and between alpha-S1 and kappa is approximately 10 and 300 kb, respectively. The beta-casein gene is oriented in the opposite direction to the alpha-S1 and kappa genes. By fluorescence in situ hybridization, the casein gene family was localized to 4q21.1.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.