Human DAO ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEJ298HU
MSPP-SEJ298HUEA 700 USD
MSPP-SEJ298HU
Human DAO ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human DAO (D-Amino Acid Oxidase). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.59 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
DAO encodes the peroxisomal enzyme D-amino acid oxidase. The enzyme is a flavoprotein which uses flavin adenine dinucleotide (FAD) as its prosthetic group. Its substrates include a wide variety of D-amino acids, but it is inactive on the naturally occurring L-amino acids. Its biological function is not known; it may act as a detoxifying agent which removes D-amino acids that accumulate during aging. In mice, it degrades D-serine, a co-agonist of the NMDA receptor. This gene may play a role in the pathophysiology of schizophrenia. The biologic role of DAMOX and DASOX has been thought to be protection against L-amino acids of fungi, bacteria and insects, or contribution to acid-base balance in the kidney.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.