Human BGN ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEJ226HU
MSPP-SEJ226HUEA 700 USD
MSPP-SEJ226HU
Human BGN ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human BGN (Biglycan). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.127 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Biglycan is a small leucine-rich repeat proteoglycan (SLRP) which is found in a variety of extracellular matrix tissues, including bone, cartilage and tendon.Biglycan consists of a protein core containing leucine-rich repeat regions and two glycosaminoglycan (GAG) chains consisting of either chondroitin sulfate (CS) or dermatan sulfate (DS), with DS being more abundant in most connective tissues. The CS/DS chains are attached at amino acids 5 and 10 in human biglycan. The composition of the GAG chains has been reported as varying according to tissue of origin. Non-glycanated forms of biglycan (no GAG chains) increase with age in human articular cartilage. The structure of biglycan core protein is highly conserved across species; over 90% homology has been reported for rat, mouse, bovine and human biglycan core proteins.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.