Mouse EPX ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEJ138MU
MSPP-SEJ138MUEA 720 USD
MSPP-SEJ138MU
Mouse EPX ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse EPX (Eosinophil Peroxidase). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.117 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Eosinophil peroxidase is a haloperoxidase enzymeencoded by the EPX gene. The enzyme is a heterodimeric 71 to 77 kDa peroxidase consisting of a heavier glycosylated chain and a lighter nonglycosylated chain. In the presence of H₂O₂ formed by the eosinophil, and either chloride or bromide ions, eosinophil peroxidase provides a potent mechanism by which eosinophils kill multicellular parasites (such as, for example, the nematode worms involved in filariasis); and also certain bacteria (such as tuberculosis bacteria). Eosinophil peroxidase is a haloperoxidase that preferentially uses bromide over chloride for this purpose, generating hypobromite (hypobromous acid). The enzyme is also capable of oxidizing thiocyanate (SCN-) and uses it as a co-substrate, with optimal concentrations occurring at about normal plasma levels.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.