Human LRRK2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEH716HU
MSPP-SEH716HUEA 700 USD
MSPP-SEH716HU
Human LRRK2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human LRRK2 (Leucine Rich Repeat Kinase 2). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 30 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
LRRK2 encodes a protein with an ankyrin repeat region, a leucine-rich repeat (LRR) domain, a kinase domain, a DFG-like motif, a RAS domain, a GTPase domain, an MLK-like domain, and a WD40 domain. The protein is present largely in the cytoplasm but also associates with the mitochondrial outer membrane. LRRK2 interacts with the C-terminal R2 RING finger domain of parkin, and parkin interacted with the COR domain of LRRK2. Expression of mutant LRRK2 induced apoptotic cell death in neuroblastoma cells and in mouse cortical neurons. Mutations in this gene have been associated with Parkinson's disease type 8. Four LRRK2 gene variants found are found in a third of Parkinson's cases, but infrequent in the general population.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.