Human SPINK5 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEH145HU
MSPP-SEH145HUEA 700 USD
MSPP-SEH145HU
Human SPINK5 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human SPINK5 (Serine Peptidase Inhibitor Kazal Type 5). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.117 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
SPINK5 encodes a multidomain serine protease inhibitor that contains 15 potential inhibitory domains. The inhibitor may play a role in skin and hair morphogenesis and anti-inflammatory and/or antimicrobial protection of mucous epithelia. Mutations may result in Netherton syndrome, a disorder characterized by ichthyosis, defective cornification, and atopy.The entire precursor protein, as deduced from the nucleotide sequence of the cloned cDNA, includes 15 potential inhibitory domains, 13 of which exhibit a Kazal-type-derived 4-cysteine-residue pattern that represents a novel protein module of serine protease inhibitor. Northern blot analysis and RT-PCR detected expression of LEKTI in thymus, vaginal epithelium, Bartholin gland, oral mucosa, tonsil, and parathyroid gland.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.