Mouse DLG4 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEG168MU
MSPP-SEG168MUEA 720 USD
MSPP-SEG168MU
Mouse DLG4 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse DLG4 (Discs, Large Homolog 4). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Disks large homolog 4 is a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with DLG2. With DLG2 it is recruited into the same NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. DLG4 is the best studied member of the MAGUK-family of PDZ domain-containing proteins. Like all MAGUK-family proteins, its basic structure includes three PDZ domains, an SH3 domain, and a guanylate kinase-like domain (GK) connected by disordered linker regions. It is almost exclusively located in the post synaptic density of neurons, and is involved in anchoring synaptic proteins.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.