Human PRDX2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEF757HU
MSPP-SEF757HUEA 700 USD
MSPP-SEF757HU
Human PRDX2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PRDX2 (Peroxiredoxin 2). The assay range is from 7.8 to 500 ng/ml (Sandwich kit) with a sensitivity of 3.1 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Peroxiredoxin-2 is a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. The crystal structure of this protein has been resolved to 2.7 angstroms. Transcript variants encoding distinct isoforms have been identified for this gene. Involved in redox regulation of the cell. Reduces peroxides with reducing equivalents provided through the thioredoxin system. It is not able to receive electrons from glutaredoxin. May play an important role in eliminating peroxides generated during metabolism.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.