Rat SIRT1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEE912RA
MSPP-SEE912RAEA 760 USD
MSPP-SEE912RA
Rat SIRT1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat SIRT1 (Sirtuin 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.054 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Sirtuin 1 is a member of the sirtuin family of proteins, homologs of the Sir2 gene in S. cerevisiae. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined, however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The protein encoded by this gene is included in class I of the sirtuin family. Sirtuin 1 is downregulated in cells that have high insulin resistance and inducing its expression increases insulin sensitivity, suggesting the molecule is associated with improving insulin sensitivity.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.