Human IL22Ra2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEE767HU
MSPP-SEE767HUEA 700 USD
MSPP-SEE767HU
Human IL22Ra2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human IL22Ra2 (Interleukin 22 Receptor Alpha 2). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.129 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
IL22Ra2 is a soluble class II cytokine receptor. This protein has been shown to specifically bind to interleukin 22 (IL22), block the interaction of IL22 with its cell surface receptor, and thus inhibit IL22 activity. This protein functions as an IL22 antagonist, and may be important in the regulation of inflammatory response. Three alternatively spliced transcript variants encoding distinct isoforms have been described.,Expressed in placenta, spleen, breast, skin and lung. Also detected in intestinal tract, testis, brain, heart and thymus. No expression found in prostate, bladder, kidney, ovary, muscle, bone marrow, liver and uterus. Isoform 1 is expressed only in placenta. Isoform 2 is expressed in placenta and breast and at lower level in spleen, skin, thymus and stomach.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.