Human NEFL ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEE038HU
MSPP-SEE038HUEA 700 USD
MSPP-SEE038HU
Human NEFL ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human NEFL (Neurofilament, Light Polypeptide). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 6.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and they functionally maintain the neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the light chain neurofilament protein. Mutations in this gene cause Charcot-Marie-Tooth disease types 1F (CMT1F) and 2E (CMT2E), disorders of the peripheral nervous system that are characterized by distinct neuropathies. A pseudogene has been identified on chromosome Y.The neurofilament subunits occupy the class IV family of intermediate filaments, and was originally thought to contain only three proteins named NF-L, NF-M and NF-H.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.