Human PGLYRP2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SED936HU
MSPP-SED936HUEA 700 USD
MSPP-SED936HU
Human PGLYRP2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PGLYRP2 (Peptidoglycan Recognition Protein 2). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
The deduced 576-amino acid precursor protein contains an N-terminal signal peptide, followed by 2 transmembrane segments and an extracellular C terminus containing PGRP domains III, II, and I. PGRPL shares 40, 33, and 32% identity with PGRPS, PGRPI-alpha, and PGRPI-beta, respectively. RNA dot blot analysis detected strong PGRPL expression in adult liver and weak expression in fetal liver. Northern blot analysis detected 2.1- and 0.8-kb transcripts expressed in adult and fetal liver. PCR also detected low expression in transverse colon, lymph nodes, heart, thymus, pancreas, descending colon, stomach, and testis. Transiently transfected COS-7 and human embryonic kidney cells expressed PGRPL as a Triton X-100-soluble membrane protein with an apparent molecular mass of 65 kD.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.