Human INSL3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SED873HU
MSPP-SED873HUEA 700 USD
MSPP-SED873HU
Human INSL3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human INSL3 (Insulin Like Protein 3). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.8 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
INSL3 encoded by this gene is an insulin like hormone produced mainly in gonadal tissues in males and females. Studies of the mouse counterpart suggest that this gene may be involved in the development of urogenital tract and female fertility. It may also act as a hormone to regulate growth and differentiation of gubernaculum, and thus mediating intra-abdominal testicular descent. The mutations in this gene may lead to, but not a frequent cause of, cryptorchidism. Expressed in prenatal and postnatal Leydig cells. Found as well in the corpus luteum, trophoblast, fetal membranes and breast.Seems to play a role in testicular function. May be a trophic hormone with a role in testicular descent in fetal life. Is a ligand for LGR8 receptor.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.