Human ABCA4 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SED620HU
MSPP-SED620HUEA 700 USD
MSPP-SED620HU
Human ABCA4 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human ABCA4 (ATP Binding Cassette Transporter A4). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.116 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
ABCA4 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. This protein is a retina-specific ABC transporter with N-retinylidene-PE as a substrate. It is expressed exclusively in retina photoreceptor cells, indicating the gene product mediates transport of an essental molecule across the photoreceptor cell membrane. Mutations in this gene are found in patients diagnosed with Stargardt disease, a form of juvenile-onset macular degeneration.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.