Mouse COX2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SED284MU
MSPP-SED284MUEA 720 USD
MSPP-SED284MU
Mouse COX2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse COX2 (Cytochrome C Oxidase Subunit II). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.114 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Cytochrome c oxidase subunit II is a human protein belongs to cytochrome c oxidase subunit II protein family. It is sometimes confused with the cyclooxygenase enzyme COX-2.Cytochrome c oxidase subunit II, abbreviated CoxII, is the second subunit of cytochrome c oxidase. Cytochrome c oxidase (EC 1.9.3.1) is an oligomeric enzymatic complex which is a component of the respiratory chain and is involved in the transfer of electrons from cytochrome c to oxygen. In eukaryotes this enzyme complex is located in the mitochondrial inner membrane; in aerobic prokaryotes it is found in the plasma membrane. The enzyme complex consists of 3-4 subunits (prokaryotes) to up to 13 polypeptides (mammals). In Leigh's disease, there may be an abnormality or deficiency of cytochrome oxidase.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.