Mouse TNNT2 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SED232MU
MSPP-SED232MUEA 720 USD
MSPP-SED232MU
Mouse TNNT2 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse TNNT2 (Troponin T Type 2 Cardiac). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.7 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Mouse TNNT2  is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Mutations in this gene have been associated with familial hypertrophic cardiomyopathy as well as with restrictive and dilated cardiomyopathy. Transcripts for this gene undergo alternative splicing that results in many tissue-specific isoforms, however, the full-length nature of some of these variants has not yet been determined. Mutations of this gene may be associated with mild or absent hypertrophy and predominant restrictive disease, with a high risk of sudden cardiac death. Advancement to dilated cardiomyopathy may be more rapid in patients with TNNT2 mutations than in those with myosin heavy chain mutations.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.