Human COL3a1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SED195HU
MSPP-SED195HUEA 700 USD
MSPP-SED195HU
Human COL3a1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human COL3a1 (Collagen Type III Alpha 1). The assay range is from 0.625 to 40 ng/ml (Sandwich kit) with a sensitivity of 0.227 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Collagen alpha-1(III) chain is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons. By fluorescence in situ hybridization, Limongi et al. (1997) concluded that the COL3A1 gene is located in chromosome band 2q32.2 and that the nebulin gene is located in band 2q24.2; the FRA2G fragile site was found to lie between the 2 genes in the 2q31 band.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.