Mouse COL6 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SED123MU
MSPP-SED123MUEA 720 USD
MSPP-SED123MU
Mouse COL6 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse COL6 (Collagen Type VI). The assay range is from 3.12 to 200 ng/ml (Sandwich kit) with a sensitivity of 1.17 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Collagen VI is a form of collagen primarily associated with the extracellular matrix of skeletal muscle.It is associated with the genes COL6A1, COL6A2, and COL6A3.Defects are associated with Bethlem myopathy and Ullrich congenital muscular dystrophy. Collagen VI is a major structural component of microfibrils. The basic structural unit of collagen VI is a heterotrimer of the alpha1(VI), alpha2(VI), and alpha3(VI) chains. The alpha2(VI) and alpha3(VI) chains are encoded by the COL6A2 and COL6A3 genes, respectively. The protein encoded by this gene is the alpha 1 subunit of type VI collagen (alpha1(VI) chain). Mutations in the genes that code for the collagen VI subunits result in the autosomal dominant disorder, Bethlem myopathy.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.