General Species GFP ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SED025GE
MSPP-SED025GEEA 816 USD
MSPP-SED025GE
General Species GFP ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting General species GFP (Green Fluorescent Protein). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.59 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
The green fluorescent protein (GFP) is a protein composed of 238 amino acid residues (26.9 kDa) that exhibits bright green fluorescence when exposed to light in the blue to ultraviolet range. Although many other marine organisms have similar green fluorescent proteins, GFP traditionally refers to the protein first isolated from the jellyfish Aequorea victoria. The GFP from A. victoria has a major excitation peak at a wavelength of 395 nm and a minor one at 475 nm. Its emission peak is at 509 nm, which is in the lower green portion of the visible spectrum.The GFP from the sea pansy has a single major excitation peak at 498 nm. In cell and molecular biology, the GFP gene is frequently used as a reporter of expression. In modified forms it has been used to make biosensors, and many animals have been created that express GFP as a proof-of-concept that a gene can be expressed throughout a given organism.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.