Human CTSB ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC964HU
MSPP-SEC964HUEA 700 USD
MSPP-SEC964HU
Human CTSB ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CTSB (Cathepsin B). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.122 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Cathepsin B is an enzymatic protein belonging to the peptidase or protease families. The protein is a lysosomal cysteine proteinase composed of a dimer of disulfide-linked heavy and light chains, both produced from a single protein precursor. It is a member of the peptidase C1 family. Cathepsin B was once suspected as a candidate protease participating in the conversion of β-amyloid precursor protein into the amyloid plaques found in Alzheimer's disease patients. However, this function is now known to be mediated by BACE1 protease. It is now thought that cathepsin B can degrade β-amyloid precursor protein into harmless fragments. Thus, it is conceivable cathepsin B may play a pivotal role in the natural defense against Alzheimer's disease.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.