Human CHGB ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC888HU
MSPP-SEC888HUEA 700 USD
MSPP-SEC888HU
Human CHGB ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CHGB (Chromogranin B). The assay range is from 0.625 to 40 ng/ml (Sandwich kit) with a sensitivity of 0.255 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Chromogranin B is a tyrosine-sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.Secretoneurin is involved in chemotaxis of monocytes and eosinophils (comparable in potency to IL8) and endothelial cells and in regulation of endothelial cell proliferation. Highest secretoneurin levels are found in anterior pituitary, followed by adrenal medulla and posterior pituitary hypothalamus (2- to 6-fold lower levels in other brain regions investigated). Secretoneurin has been shown to have potent angiogenic activity in vivo in mouse cornea model and in vitro in a 3-dimensional gel.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.