Human SOST ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC864HU
MSPP-SEC864HUEA 700 USD
MSPP-SEC864HU
Human SOST ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human SOST (Sclerostin). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.131 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Sclerostin is a secreted glycoprotein with a C-terminal cysteine knot-like (CTCK) domain and sequence similarity to the DAN (differential screening-selected gene aberrative in neuroblastoma) family of bone morphogenetic protein (BMP) antagonists. Loss-of-function mutations in this gene are associated with an autosomal-recessive disorder, sclerosteosis, which causes progressive bone overgrowth. A deletion downstream of this gene, which causes reduced sclerostin expression, is associated with a milder form of the disorder called van Buchem disease.Sclerostin is produced by the osteocyte. These cells originate from osteoblasts, the cells that form the bone matrix. Sclerostin has an inhibitory effect on the life time of the osteoblast. Osteoblasts are self limiting by a negative feedback system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.