Human TSG101 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC814HU
MSPP-SEC814HUEA 700 USD
MSPP-SEC814HU
Human TSG101 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human TSG101 (Tumor Susceptibility Gene 101). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.117 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Tumor susceptibility gene 101, also known as TSG101, is a protein belongs to a group of apparently inactive homologs of ubiquitin-conjugating enzymes. The gene product contains a coiled-coil domain that interacts with stathmin, a cytosolic phosphoprotein implicated in tumorigenesis. The protein may play a role in cell growth and differentiation and act as a negative growth regulator. in vitro steady-state expression of this tumor susceptibility gene appears to be important for maintenance of genomic stability and cell cycle regulation. Mutations and alternative splicing in this gene occur in high frequency in breast cancer and suggest that defects occur during breast cancer tumorigenesis and/or progression.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.