Human PAM ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC744HU
MSPP-SEC744HUEA 700 USD
MSPP-SEC744HU
Human PAM ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human PAM (Peptidylglycine Alpha Amidating Monooxygenase). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.9 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Peptidyl-glycine alpha-amidating monooxygenase is a multifunctional protein. It has two enzymatically active domains with catalytic activities - peptidylglycine alpha-hydroxylating monooxygenase (PHM) and peptidyl-alpha-hydroxyglycine alpha-amidating lyase (PAL). These catalytic domains work sequentially to catalyze neuroendocrine peptides to active alpha-amidated products. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene but some of their full length sequences are not yet known.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.