Rat VNN1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC598RA
MSPP-SEC598RAEA 760 USD
MSPP-SEC598RA
Rat VNN1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Rat VNN1 (Vanin 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.062 ng/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Pantetheinase is a member of the Vanin family of proteins which share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. This protein, like its mouse homolog, is likely a GPI-anchored cell surface molecule. The mouse protein is expressed by the perivascular thymic stromal cells and regulates migration of T-cell progenitors to the thymus. This gene lies in close proximity to, and in same transcriptional orientation as two other vanin genes on chromosome 6q23 to q24.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.