Human LOR ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC568HU
MSPP-SEC568HUEA 700 USD
MSPP-SEC568HU
Human LOR ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human LOR (Loricrin). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.058 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Loricrin is a major protein component of the cornified cell envelope found in terminally differentiated epidermal cells. Mutations in this gene are associated with Vohwinkel's syndrome and progressive symmetric erythrokeratoderma, both inherited skin diseases. There were multiple sequence variants within these 2 size class alleles due to various deletions of 12 bp (4 amino acids) in the major loop of this glycine loop domain. By use of a specific loricrin antibody in immunogold electron microscopy, they showed that loricrin appears initially in the granular layer of human epidermis and forms composite keratohyalin granules with profilaggrin, but localizes to the cell periphery (cell envelope) of fully differentiated stratum corneum cells.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.