Mouse FPN ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC489MU
MSPP-SEC489MUEA 720 USD
MSPP-SEC489MU
Mouse FPN ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse FPN (Ferroportin). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.056 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Ferroportin is a transmembrane protein that transports iron from the inside of a cell to the outside of it. Recent research suggests that ferroportin is inhibited by hepcidin, which therefore is the 'master regulator' of human iron metabolism. Hepcidin binds to ferroportin, and results in the internalisation of ferroportin within the cell, followed by degradation by the proteasome. This results in retention of iron within the cell, and a reduction in iron levels within the plasma. This is part of the mechanism that causes anaemia of chronic disease; hepcidin is released from the liver in response to inflammatory cytokines, namely interleukin-6, which results in an increased hepcidin concentration and a consequent decrease in plasma iron levels.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.