Human FGg ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC477HU
MSPP-SEC477HUEA 700 USD
MSPP-SEC477HU
Human FGg ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human FGg (Fibrinogen Gamma). The assay range is from 0.625 to 40 ng/ml (Sandwich kit) with a sensitivity of 0.264 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Fibrinogen gamma chain is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.