Human NUMA1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC332HU
MSPP-SEC332HUEA 700 USD
MSPP-SEC332HU
Human NUMA1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human NUMA1 (Nuclear Mitotic Apparatus Protein 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.055 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
The NUMA protein was one of the first to be described as a cell cycle-related protein based on a distinct immunofluorescent staining pattern: in interphase, NUMA is present throughout the nucleus, and in mitosis, it localizes to the spindle apparatus (Lydersen and Pettijohn, 1980). Some patients with autoimmune disease have antibodies directed against the NUMA protein. The full-length NUMA cDNA (Compton et al., 1992; Yang et al., 1992) predicts a protein with a calculated molecular mass of about 237 kD, containing 2 globular domains separated by a coiled-coil domain of 1,485 amino acids. Wong et al. (2006) stated that the coiled-coil region of NUMA can form parallel coiled-coil dimers. By fluorescence in situ hybridization, Sparks et al. (1993) demonstrated that the NUMA1 gene is present in single copy and located on chromosome 11q13.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.