Human TJP1 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC262HU
MSPP-SEC262HUEA 700 USD
MSPP-SEC262HU
Human TJP1 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human TJP1 (Tight Junction Protein 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Tight junction protein ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. The encoded protein may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for this gene. Tight junction (zonula occludens) protein-1 (TJP1), also referred to as ZO-1, is a 200 kDa protein located on a cytoplasmic membrane surface of vertebrate intercellular tight junctions. Willott et al., (1993) isolated a full-length cDNA sequence for human TJP1. Although the function of TJP1 is unknown, the cDNA sequence predicted a multi-domain signaling protein homologous to the product of the 'discs large-1' tumor suppressor gene of Drosophila  and several other membrane-associated proteins in mammals.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.