Human TF ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC036HU
MSPP-SEC036HUEA 419 USD
MSPP-SEC036HU
Human TF ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human TF (Transferrin). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.74 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%
Transferrin is a blood plasma protein for iron ion delivery that, in humans, is encoded by the TF gene. Transferrin is a glycoprotein that binds iron very tightly but reversibly. Although iron bound to transferrin is less than 0.1% (4 mg) of the total body iron, it is the most important iron pool, with the highest rate of turnover (25 mg/24 h). Transferrin has a molecular weight of around 80 kDa and contains 2 specific high-affinity Fe (III) binding sites. The affinity of transferrin for Fe (III) is extremely high (1023 M-1 at pH 7.4) [citation needed] but decreases progressively with decreasing pH below neutrality.In humans, transferrin consists of a poly peptide chain containing 679 amino acids. It is a complex composed of alpha helices and beta sheets to form two domains.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.