Human CDH17 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEC002HU
MSPP-SEC002HUEA 700 USD
MSPP-SEC002HU
Human CDH17 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human CDH17 (Cadherin 17). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 13.1 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Cadherin-17 is a member of the cadherin superfamily, genes encoding calcium-dependent, membrane-associated glycoproteins. The encoded protein is cadherin-like, consisting of an extracellular region, containing 7 cadherin domains, and a transmembrane region but lacking the conserved cytoplasmic domain. The protein is a component of the gastrointestinal tract and pancreatic ducts, acting as an intestinal proton-dependent peptide transporter in the first step in oral absorption of many medically important peptide-based drugs. The protein may also play a role in the morphological organization of liver and intestine.Alternative splicing results in multiple transcript variants.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.