Human HJV ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB995HU
MSPP-SEB995HUEA 700 USD
MSPP-SEB995HU
Human HJV ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human HJV (Hemojuvelin). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 12.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Hemojuvelin is transcribed from a gene of 4265 bp into a full-length transcript with 5 spliced isoforms. The putative full-length protein from the longest transcript is 426 amino acids. Hemojuvelin contains multiple protein motifs consistent with a function as a membrane-bound receptor or secreted polypeptide hormone. Intraperitoneal injection of BMP6 in mice caused increased hepatic hepcidin mRNA expression and reduced serum iron and transferrin saturation in a dose-dependent manner. Conversely, inhibition of endogenous Bmp6 in mice reduced hepcidin expression and increased serum iron. Andriopoulos et al. (2009) concluded that BMP6 is an HJV ligand and an endogenous regulator of hepcidin expression and iron metabolism.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.