Human KLK7 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB910HU
MSPP-SEB910HUEA 560 USD
MSPP-SEB910HU
Human KLK7 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Human KLK7 (Kallikrein 7). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.059 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Kallikrein-related peptidase 7, also known as KLK7, Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its encoded enzyme is thought to be involved in the proteolysis of intercellular cohesive structures preceding desquamation, which is the shedding of the outermost layer of the epidermis. Alternative splicing of this gene results in two transcript variants encoding the same protein.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.