Mouse APOC3 ELISA Kit
Supplier: CLOUD-CLONE CORP MS
SEB890MU
MSPP-SEB890MUEA 684 USD
MSPP-SEB890MU
Mouse APOC3 ELISA Kit
Assays ELISAs
This assay has high sensitivity and excellent specificity for detecting Mouse APOC3 (Apolipoprotein C3). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.37 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%
Apolipoprotein C-III is a very low density lipoprotein (VLDL) protein. APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia.Lately, two novel susceptibility haplotypes i.e., P2-S2-X1 and P1-S2-X1 have been discovered in ApoAI-CIII-AIV gene cluster on chromosme 11q23, who confer approximately threefold higher risk of coronary heart disease in normal as well as non-insulin diabetes mellitus.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.